2-Methyl-3-butyne-2-ol ≥98.0%

Supplier: TCI America

M0180-25ML M0180-500ML
TCM0180-025MLEA 21 USD
TCM0180-025ML TCM0180-500ML
2-Methyl-3-butyne-2-ol ≥98.0%
2-Methyl-3-butyne-2-ol

CAS Number: 115-19-5
MDL Number: MFCD00004467
Molecular Formula: C5H8O
Molecular Weight: 84.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Flash Point (°C): 22
Specific Gravity (20/20): 0.86


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Click Chemistry
Other Product Names / Synonyms: 3-Methyl-1-butyn-3-ol

Formula: C₅H₈O
Boiling Pt: 103 °C
Melting Pt: 3 °C
Density: 0.86 (20 °C)
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00004467
CAS Number: 115-19-5
EINECS: 204-070-5
UN: 1993
ADR: 3,II
Merck Index: 12,06113
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8d50fdfa49d3767e7aea2879d43b458f11a1b9b&vid=a8d50fdfa49d3767e7aea2879d43b458f11a1b9b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR