2-Methyl-3-buten-2-ol ≥97.0%

Supplier: TCI America

Synonyms: 3-Hydroxy-3-methyl-1-butene

M0178-500ML M0178-25ML
TCM0178-500MLEA 226 USD
TCM0178-500ML TCM0178-25ML
2-Methyl-3-buten-2-ol ≥97.0%
2-Methyl-3-buten-2-ol

CAS Number: 115-18-4
MDL Number: MFCD00004470
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Melting point (°C): -28
Flash Point (°C): 19
Specific Gravity (20/20): 0.82


Product Categories: Bioscience, Hemiterpenes
Other Product Names / Synonyms: 3-Hydroxy-3-methyl-1-butene

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 96 °C
Melting Pt: –28 °C
Density: 0.82 (20 °C)
Flash Pt: 19 °C
Storage Temperature: Ambient
MDL Number: MFCD00004470
CAS Number: 115-18-4
EINECS: 204-068-4
UN: 1987
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86ab09fb78d954aceee376d00b8c72529a6c393f&vid=86ab09fb78d954aceee376d00b8c72529a6c393f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR