2-Methylbutane ≥98.0%

Supplier: TCI America
Danger

Synonyms: iso-Pentane

M0167-25ML M0167-500ML
TCM0167-25MLEA 24.19 USD
TCM0167-25ML TCM0167-500ML
2-Methylbutane ≥98.0%
2-Methylbutane

CAS Number: 78-78-4
MDL Number: MFCD00009338
Molecular Formula: C5H12
Molecular Weight: 72.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 31
Flash Point (°C): -57
Specific Gravity (20/20): 0.62


Other Product Names / Synonyms: Isopentane

Formula: CH₃CH₂CH(CH₃)₂
MW: 72.15 g/mol
Boiling Pt: 28
Melting Pt: -160
Density: 0.62 (20 °C)
Flash Pt: –57 °C
Storage Temperature: Ambient
MDL Number: MFCD00009338
CAS Number: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffa9dacd078716b6de2e96d98f8b77f716b3e104&vid=8c0df4468ce362cac37edbcdb3ac3ae0f3972b03&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR