(±)-α-Methylbenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: DL-1-Phenylethylamine, (±)-1-Phenylethylamine, (±)-α-Phenylethylamine

M0165-25ML M0165-100ML
TCM0165-25MLEA 38.95 USD
TCM0165-25ML TCM0165-100ML
(±)-α-Methylbenzylamine ≥98.0% (by GC, titration analysis)
(±)-α-Methylbenzylamine

CAS Number: 618-36-0
MDL Number: MFCD00008069
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 185
Melting point (°C): -65
Flash Point (°C): 69
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: DL-alpha-Methylbenzylamine

Formula: C₆H₅CH(NH₂)CH₃
MW: 121.18 g/mol
Boiling Pt: 185 °C
Melting Pt: –65 °C
Density: 0.96 (20 °C)
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00008069
CAS Number: 618-36-0
EINECS: 210-545-8
UN: 2735
ADR: 8,II
Merck Index: 12,06106
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e5ea2077054880bc8e2c239d7ac54bece2d2ef3&vid=7e5ea2077054880bc8e2c239d7ac54bece2d2ef3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR