Benzyl(methyl)amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: N-Benzylmethylamine, N-Methyl-1-phenylmethanamine, N-Methylbenzylamine

M0164-500ML M0164-25ML
TCM0164-500MLEA 83.35 USD
TCM0164-500ML TCM0164-25ML
Benzyl(methyl)amine ≥98.0% (by GC, titration analysis)
Benzyl(methyl)amine

CAS Number: 103-67-3
MDL Number: MFCD00008289
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 186
Melting point (°C): -55
Flash Point (°C): 77
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: N-Benzylmethylamine, omega-Methylaminotoluene

Formula: C₆H₅CH₂NHCH₃
MW: 121.18 g/mol
Boiling Pt: 186 °C
Melting Pt: –55 °C
Density: 0.94 (20 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00008289
CAS Number: 103-67-3
EINECS: 203-133-4
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01f4194524f267892b0ce5e72bb17a69d2ddb0a6&vid=01f4194524f267892b0ce5e72bb17a69d2ddb0a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR