(±)-1-Phenylethanol ≥98.0%

Supplier: TCI America
Danger

Synonyms: (±)-sec-Phenethyl alcohol, Styrene alcohol, (±)-α-Methylbenzyl alcohol, (±)-1-Phenylethyl alcohol, 1-Phenylethanol, Methyl phenyl carbinol, α-Methylbenzyl alcohol, Styrallyl alcohol

M0163-500ML M0163-25ML
TCM0163-500MLEA 100 USD
TCM0163-500ML TCM0163-25ML
(±)-1-Phenylethanol ≥98.0%
(±)-1-Phenylethanol

CAS Number: 98-85-1
MDL Number: MFCD00004508
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 94
Freezing point (°C): 20
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: (+/-)-alpha-Methylbenzyl Alcohol, (+/-)-1-Phenylethanol, (+/-)-sec-Phenethyl Alcohol

Formula: C₆H₅CH(OH)CH₃
MW: 122.17 g/mol
Boiling Pt: 205 °C
Melting Pt: 20 °C
Density: 1.01 (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00004508
CAS Number: 98-85-1
EINECS: 202-707-1
UN: 2937
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7a48732b0152ae5ce883ef49494a91b3fad9e0f&vid=b7a48732b0152ae5ce883ef49494a91b3fad9e0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR