N-Methylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

M0147-500G M0147-25G
TCM0147-500GEA 44 USD
TCM0147-500G TCM0147-25G
N-Methylaniline ≥98.0% (by GC, titration analysis)
N-Methylaniline

CAS Number: 100-61-8
MDL Number: MFCD00008283
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 196
Melting point (°C): -57
Flash Point (°C): 83
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: N-Monomethylaniline

Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 196 °C
Melting Pt: -57 °C
Density: 0.99 (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00008283
CAS Number: 100-61-8
EINECS: 202-870-9
UN: 2294
ADR: 6.1,III
Merck Index: 12,06098
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=978691bad2b138bcad1fbb38c1a9261c8c53241f&vid=7546da49c5448b0f014c8b1f6a76ae3ced97f402&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR