4-(Methylamino)phenol sulfate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
M0145-25G
M0145-500G
TCM0145-025GEA
43.6
USD
TCM0145-025G
TCM0145-500G
4-(Methylamino)phenol sulfate ≥98.0% (by HPLC, total nitrogen)
4-(Methylamino)phenol sulfate
CAS Number: 55-55-0
MDL Number: MFCD00013140
Molecular Formula: C7H9NO
Molecular Weight: 344.38
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 260
Formula:
C₁₄H₂₀N₂O₆S Boiling Pt: 227 °C (1013 hPa) Melting Pt: 260 °C Density: 1.434 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013140 CAS Number: 55-55-0 EINECS: 200-237-1 UN: 2811 ADR: 6.1,III Merck Index: 13,06046 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...