2-Methylaminoethanol ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: N-Methyl-2-ethanolamin, N-Methyl-2-aminoethanol, 2-(Methylamino)ethanol

M0144-500ML M0144-25ML
TCM0144-500MLEA 51.76 USD
TCM0144-500ML TCM0144-25ML
2-Methylaminoethanol ≥99.0% (by GC, titration analysis)
2-Methylaminoethanol

CAS Number: 109-83-1
MDL Number: MFCD00002839
Molecular Formula: C3H9NO
Molecular Weight: 75.11
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 156
Melting point (°C): -5
Flash Point (°C): 73
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: N-Methylethanolamine

Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 156 °C
Melting Pt: –5 °C
Density: 0.94 (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002839
CAS Number: 109-83-1
EINECS: 203-710-0
Merck Index: 12,06096
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3b32436b246d3f0630055ea17f1d37b2d8912d&vid=db3b32436b246d3f0630055ea17f1d37b2d8912d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR