1-Amino-3-methoxypropane ≥99.0%

Supplier: TCI America

M0127-500ML M0127-25ML
TCM0127-500MLEA 47.1 USD
TCM0127-500ML TCM0127-25ML
1-Amino-3-methoxypropane ≥99.0%
1-Amino-3-methoxypropane

CAS Number: 5332-73-0
MDL Number: MFCD00014831
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 32
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: 3-Aminopropyl Methyl Ether, Propanolamine Methyl Ether

Formula: C₄H₁₁NO
Boiling Pt: 120 °C
Melting Pt: < –70 °C
Density: 0.87 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00014831
CAS Number: 5332-73-0
EINECS: 226-241-3
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36382e1e67ae1ac9e149d5cd87759ded06aac358&vid=36382e1e67ae1ac9e149d5cd87759ded06aac358&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR