(±)-1-Methoxy-2-propanol ≥98.0%
Supplier: TCI America
|
Danger
|
CAS Number: 107-98-2
MDL Number: MFCD00004537
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Melting point (°C): -96
Flash Point (°C): 32
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: Propylene Glycol 1-Monomethyl Ether
Formula:
CH₃CH(OH)CH₂OCH₃ Boiling Pt: 120 °C Melting Pt: -96 °C Density: 0.92 (20 °C) Flash Pt: 32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004537 CAS Number: 107-98-2 EINECS: 203-539-1 UN: 3092 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...