3-Methoxyphenol ≥98.0%

Supplier: TCI America
Warning

M0122-500ML M0122-25ML M0122-100ML
TCM0122-500MLEA 333 USD
TCM0122-500ML TCM0122-025ML TCM0122-100ML
3-Methoxyphenol ≥98.0%
3-Methoxyphenol

CAS Number: 150-19-6
MDL Number: MFCD00002267
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 244
Flash Point (°C): 114
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: 3-Hydroxyanisole, Resorcinol Monomethyl Ether

Formula: C₇H₈O₂
Boiling Pt: 244 °C
Melting Pt: –2 °C
Density: 1.15 (20 °C)
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00002267
CAS Number: 150-19-6
EINECS: 205-754-6
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0ef01b8db74e92696af81c8091dac93a0c86529&vid=d95f24e3d08c214cc54f9f3f59e8a794779c0cc9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR