2-Nitro-p-anisidine ≥98.0%

Supplier: TCI America
Danger

Synonyms: 4-Amino-3-nitroanisole, 4-Methoxy-2-nitroaniline

M0119-25G M0119-100G
TCM0119-25GEA 16.28 USD
TCM0119-25G TCM0119-100G
2-Nitro-p-anisidine ≥98.0%
2-Nitro-p-anisidine

CAS Number: 96-96-8
MDL Number: MFCD00007152
Molecular Formula: C7H8N2O3
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 124


Other Product Names / Synonyms: 1-Amino-4-methoxy-2-nitrobenzene, 2-Nitro-p-anisidine

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 124 °C
Flash Pt: >200 °C
MDL Number: MFCD00007152
CAS Number: 96-96-8
EINECS: 202-547-2
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c65ea93b4625e221c55fc66ffd8ddeb924c77048&vid=c65ea93b4625e221c55fc66ffd8ddeb924c77048&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR