1-Methoxynaphthalene ≥98.0%

Supplier: TCI America

M0116-500G M0116-25G
TCM0116-500GEA 325 USD
TCM0116-500G TCM0116-025G
1-Methoxynaphthalene ≥98.0%

CAS Number: 2216-69-5
MDL Number: MFCD00003924
Molecular Formula: C11H10O
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 271
Melting point (°C): 6
Flash Point (°C): 112
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: Methyl 1-Naphthyl Ether

Formula: C₁₁H₁₀O
Boiling Pt: 271 °C
Melting Pt: 6 °C
Density: 1.10 (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00003924
CAS Number: 2216-69-5
EINECS: 218-696-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8096099352a13a8ced925d6aaafd87f6941786ec&vid=8096099352a13a8ced925d6aaafd87f6941786ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR