3-Methoxy-1-butanol ≥98.0%

Supplier: TCI America

Synonyms: 3-Methoxybutan-1-ol

M0109-25ML M0109-500ML
TCM0109-25MLEA 21 USD
TCM0109-25ML TCM0109-500ML
3-Methoxy-1-butanol ≥98.0%
3-Methoxy-1-butanol

CAS Number: 2517-43-3
MDL Number: MFCD00002931
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 64
Specific Gravity (20/20): 0.92

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 158 °C
Melting Pt: -85 °C
Density: 0.92 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00002931
CAS Number: 2517-43-3
EINECS: 219-741-8
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b7644b6050a8a56e2913a490db3eb520e85f701&vid=0b7644b6050a8a56e2913a490db3eb520e85f701&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR