2-Anisoyl chloride ≥98.0%

Supplier: TCI America
Danger

Synonyms: 2-Methoxybenzoyl chloride, 2-Methoxybenzene-1-carbonyl chloride

M0108-500G M0108-25G M0108-100G
TCM0108-500GEA 990 USD
TCM0108-500G TCM0108-25G TCM0108-100G
2-Anisoyl chloride ≥98.0%
2-Anisoyl chloride

CAS Number: 21615-34-9
MDL Number: MFCD00000664
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 84
Specific Gravity (20/20): 1.27


Other Product Names / Synonyms: o-Anisoyl Chloride

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 137
Density: 1.27 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00000664
CAS Number: 21615-34-9
EINECS: 244-477-5
UN: 1729
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eec14c496ccfa933e77989d0f80dc4de6acb6e9&vid=6eec14c496ccfa933e77989d0f80dc4de6acb6e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR