Methacrylic acid ≥99.0% stabilized

Supplier: TCI America
Danger

Synonyms: 2-Methylpropenoic acid

Stabilization: Stabilised with Stabilised

M0079-25G M0079-500G
TCM0079-25GEA 23.48 USD
TCM0079-25G TCM0079-500G
Methacrylic acid ≥99.0% stabilized
Methacrylic acid

(stabilized with MEHQ)
CAS Number: 79-41-4
MDL Number: MFCD00002651
Molecular Formula: C4H6O2
Molecular Weight: 86.09
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 77
Freezing point (°C): 15
Specific Gravity (20/20): 1.02

Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 160 °C
Melting Pt: 15 °C
Density: 1.02 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00002651
CAS Number: 79-41-4
EINECS: 201-204-4
UN: 2531
ADR: 8,II
Merck Index: 13,05967
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3994515d4a04cc85f59e28fcaa566d3032bd5a42&vid=3994515d4a04cc85f59e28fcaa566d3032bd5a42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR