(±)-Mercaptosuccinic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
CAS Number: 70-49-5
MDL Number: MFCD00004860
Molecular Formula: C4H6O4S
Molecular Weight: 150.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 152
Other Product Names / Synonyms: Mercaptosuccinic Acid
Formula:
C₄H₆O₄S Boiling Pt: 322 °C (1013 hPa) Melting Pt: 152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004860 CAS Number: 70-49-5 EINECS: 200-736-4 Merck Index: 12,09479 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...