2-Mercaptobenzothiazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT

M0055-500G M0055-25G
TCM0055-500GEA 47 USD
TCM0055-500G TCM0055-25G
2-Mercaptobenzothiazole ≥98.0% (by HPLC, titration analysis)
2-Mercaptobenzothiazole

CAS Number: 149-30-4
MDL Number: MFCD00005781
Molecular Formula: C7H5NS2
Molecular Weight: 167.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 180
Flash Point (°C): 200
Lambda max.: 325 nm (MeOH)

Formula: C₇H₅NS₂
MW: 167.26 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 180 °C
Density: 1.42 g/cm³ (20 °C)
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00005781
CAS Number: 149-30-4
EINECS: 205-736-8
UN: 3077
ADR: 9,III
Merck Index: 13,05893
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36bae1e7599a9a34669875ab9fbf318a4b71a4fa&vid=36bae1e7599a9a34669875ab9fbf318a4b71a4fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR