Thiosalicylic acid ≥90.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Thiobenzoic acid, 2-Mercaptobenzoic acid

M0054-25G M0054-500G
TCM0054-025GEA 30.3 USD
TCM0054-025G TCM0054-500G
Thiosalicylic acid ≥90.0% (by titrimetric analysis)
Thiosalicylic acid

CAS Number: 147-93-3
MDL Number: MFCD00004836
Molecular Formula: C7H6O2S
Molecular Weight: 154.18
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Melting point (°C): 167


Other Product Names / Synonyms: 2-Mercaptobenzoic Acid

Formula: C₇H₆O₂S
MW: 154.19 g/mol
Boiling Pt: 230 °C (0.02 -rr)
Melting Pt: 167 °C
Density: 1.489 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004836
CAS Number: 147-93-3
EINECS: 205-704-3
UN: 3335
ADR: 9,
Merck Index: 12,09498
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37ab6030eb92adcd4267f53cc4e26955c4230aba&vid=5d2633365a28188b60a167631d9033bfaf0d02dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR