Maleic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Warning

M0006-500G M0006-25G
TCM0006-500GEA 47.78 USD
TCM0006-500G TCM0006-25G
Maleic acid ≥99.0% (by titrimetric analysis)
Maleic acid

CAS Number: 110-16-7
MDL Number: MFCD00063177
Molecular Formula: C4H4O4
Molecular Weight: 116.07
Purity/Analysis Method: >99.0% (T)
Form: Crystalline Powder
Melting point (°C): 131
Flash Point (°C): 138


Other Product Names / Synonyms: cis-2-Butenedioic Acid

Formula: C₄H₄O₄
MW: 116.07 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 131 °C
Density: 1.59 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7
EINECS: 203-742-5
UN: 3261
ADR: 8,III
Merck Index: 13,05726
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ea35067ba14d1258c4b279c2a1a5bd3ae0ee158&vid=7ea35067ba14d1258c4b279c2a1a5bd3ae0ee158&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR