Lithium carbonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

L0224-25G L0224-500G
TCL0224-25GEA 26.48 USD
TCL0224-25G TCL0224-500G
Lithium carbonate ≥98.0% (by titrimetric analysis)
Lithium carbonate

CAS Number: 554-13-2
MDL Number: MFCD00011084
Molecular Formula: CLi2O3
Molecular Weight: 73.89
Purity/Analysis Method: >98.0% (T)
Form: Powder
Melting point (°C): 723

Formula: Li₂CO₃
Melting Pt: 723 °C
Density: 2.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR