Methyl (S)-(-)-lactate ≥98.0%

Supplier: TCI America
Danger

Synonyms: (S)-Methyl 2-hydroxypropanoate, Methyl (S)-(-)-2-hydroxypropionate, Sodium carboxymethyl cellulose

L0163-25G L0163-500G L0163-100G
TCL0163-025GEA 15.67 USD
TCL0163-025G TCL0163-500G TCL0163-100G
Methyl (S)-(-)-lactate ≥98.0%
Methyl (S)-(-)-lactate

CAS Number: 27871-49-4
MDL Number: MFCD00064265
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 49
Specific Gravity (20/20): 1.10
Specific rotation [a]20/D: -8.2 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: L-(-)-Lactic Acid Methyl Ester

Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 140 °C
Melting Pt: –44 °C
Density: 1.10 (20 °C)
Flash Pt: 49 °C
MDL Number: MFCD00064265
CAS Number: 27871-49-4
EINECS: 248-704-9
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=503f64672f1415c656114848bb81df5281166a32&vid=503f64672f1415c656114848bb81df5281166a32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR