Lidocaine ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

L0156-25G L0156-500G L0156-100G
TCL0156-025GEA 69.02 USD
TCL0156-025G TCL0156-500G TCL0156-100G
Lidocaine ≥99.0% (by HPLC, titration analysis)
Lidocaine

CAS Number: 137-58-6
MDL Number: MFCD00026733
Molecular Formula: C14H22N2O
Molecular Weight: 234.34
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 182
Melting point (°C): 69


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2-Diethylamino-N-(2,6-dimethylphenyl)acetamide

Formula: C₁₄H₂₂N₂O
Boiling Pt: 182 °C
Melting Pt: 69 °C
MDL Number: MFCD00026733
CAS Number: 137-58-6
EINECS: 205-302-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0d86ad5c057bb82881fa68cad9cc0a023427b1a&vid=e0d86ad5c057bb82881fa68cad9cc0a023427b1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR