2,4-Lutidine ≥98.0%

Supplier: TCI America
Danger

Synonyms: 2,4-Dimethylpyridine

L0085-25ML L0085-500ML
TCL0085-025MLEA 45 USD
TCL0085-025ML TCL0085-500ML
2,4-Lutidine ≥98.0%
2,4-Lutidine

CAS Number: 108-47-4
MDL Number: MFCD00006337
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Melting point (°C): -64
Flash Point (°C): 37
Specific Gravity (20/20): 0.93


Product Categories: Synthetic Organic Chemistry, Ligands, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: 2,4-Dimethylpyridine

Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 157 °C
Melting Pt: -64 °C
Density: 0.93 (20 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00006337
CAS Number: 108-47-4
EINECS: 203-586-8
UN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f761f6047f7b983c9d223e2df24bccaa8bd7ec91&vid=f761f6047f7b983c9d223e2df24bccaa8bd7ec91&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR