2,6-Lutidine ≥98.0%
Supplier: TCI America
Synonyms:
2,6-Dimethylpyridine
L0067-500ML
L0067-25ML
TCL0067-500MLEA
140.22
USD
TCL0067-500ML
TCL0067-25ML
2,6-Lutidine ≥98.0%
2,6-Lutidine
CAS Number: 108-48-5
MDL Number: MFCD00006345
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Melting point (°C): -6
Flash Point (°C): 33
Specific Gravity (20/20): 0.92
Product Categories: Synthetic Organic Chemistry, Ligands, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: 2,6-Dimethylpyridine
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 144 °C Melting Pt: –6 °C Density: 0.92 (20 °C) Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006345 CAS Number: 108-48-5 EINECS: 203-587-3 UN: 1993 ADR: 3,III Merck Index: 13,05636 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...