Levulinic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Oxovaleric acid, 4-Ketovaleric acid, 4-Oxopentanoic acid

L0042-500G L0042-25G
TCL0042-500GEA 68.38 USD
TCL0042-500G TCL0042-25G
Levulinic acid ≥97.0% (by GC, titration analysis)
Levulinic acid

CAS Number: 123-76-2
MDL Number: MFCD00002796
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 246
Flash Point (°C): 137
Freezing point (°C): 29


Product Categories: Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: 3-Acetylpropionic Acid, 4-Oxovaleric Acid

Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 246 °C
Melting Pt: 26 - 29 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: 137 °C
MDL Number: MFCD00002796
CAS Number: 123-76-2
EINECS: 204-649-2
UN: 3261
ADR: 8,III
Merck Index: 13,05492

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR