Lead tetraacetate ≥96.0% (by titrimetric analysis) (contains Acetic Acid)

Supplier: TCI America
Danger

L0021-500G L0021-25G
TCL0021-500GEA 583 USD
TCL0021-500G TCL0021-25G
Lead tetraacetate ≥96.0% (by titrimetric analysis) (contains Acetic Acid)
Lead tetraacetate

CAS Number: 546-67-8
MDL Number: MFCD00008693
Molecular Formula: C2H4O2
Molecular Weight: 443.38
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 180
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Oxidation, Reagents for the Generation of Arynes (Aryne Precursors)
Other Product Names / Synonyms: LTA

Formula: (H₃CCOO)₄Pb
Melting Pt: 180 °C
Density: ~ 2.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008693
CAS Number: 546-67-8
EINECS: 208-908-0
UN: 1616
ADR: 6.1,III
Merck Index: 13,05442
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d2baa2454f3fed03ec227df6aad5378dd3cff5c&vid=99bc5581abd632c1308d35bbc90594d65816329b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR