Kojic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone
K0010-25G
K0010-500G
TCK0010-025GEA
69
USD
TCK0010-025G
TCK0010-500G
Kojic acid ≥98.0% (by titrimetric analysis)
Kojic acid
CAS Number: 501-30-4
MDL Number: MFCD00006580
Molecular Formula: C6H6O4
Molecular Weight: 142.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 154
Lambda max.: 269 nm (CHCl3)
Product Categories: Bioscience, Cosmetic Ingredients for Research and Experimental Use
Other Product Names / Synonyms: 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
Formula:
C₆H₆O₄ MW: 142.11 g/mol Boiling Pt: 323 °C (1013 hPa) Melting Pt: 154 °C Density: 1.58 g/cm³ (20 °C) |
MDL Number:
MFCD00006580 CAS Number: 501-30-4 EINECS: 207-922-4 Merck Index: 13,05334 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...