3-Oxoglutaric acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America

K0006-25G K0006-250G
TCK0006-025GEA 58.6 USD
TCK0006-025G TCK0006-250G
3-Oxoglutaric acid ≥95.0% (by titrimetric analysis)
3-Oxoglutaric acid

CAS Number: 542-05-2
MDL Number: MFCD00002711
Molecular Formula: C5H6O5
Molecular Weight: 146.10
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 126
Storage Temperature: <0°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3-Oxoglutaric Acid, 3-Oxopentanedioic Acid

Formula: C₅H₆O₅
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 126 °C
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00002711
CAS Number: 542-05-2
EINECS: 208-797-9
Merck Index: 13,00069
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3eda42c8ddf65b46f57178da9afe670c88c8e26&vid=b3eda42c8ddf65b46f57178da9afe670c88c8e26&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR