2-Oxoglutaric acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Danger

K0005-25G K0005-500G K0005-100G
TCK0005-025GEA 53.92 USD
TCK0005-025G TCK0005-500G TCK0005-100G
2-Oxoglutaric acid ≥99.0% (by titrimetric analysis)
2-Oxoglutaric acid

CAS Number: 328-50-7
MDL Number: MFCD00004165
Molecular Formula: C5H6O5
Molecular Weight: 146.10
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 116


Other Product Names / Synonyms: 2-Ketoglutaric Acid

Formula: HOOCCH₂CH₂COCOOH
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004165
CAS Number: 328-50-7
EINECS: 206-330-3
Merck Index: 13,05320
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3777d5abbf08973c33da7825f35dfef3c172697&vid=a3777d5abbf08973c33da7825f35dfef3c172697&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR