2-Chlorophenyl isothiocyanate ≥95.0%

Supplier: TCI America

Synonyms: 1-Chloro-2-isothiocyanatobenzene

I0534-25G I0534-5G
TCI0534-25GEA 39 USD
TCI0534-25G TCI0534-5G
2-Chlorophenyl isothiocyanate ≥95.0%
2-Chlorophenyl isothiocyanate

CAS Number: 2740-81-0
MDL Number: MFCD00004801
Molecular Formula: C7H4ClNS
Molecular Weight: 169.63
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 262
Flash Point (°C): 110
Specific Gravity (20/20): 1.30


Other Product Names / Synonyms: Isothiocyanic Acid 2-Chlorophenyl Ester

Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 262 °C
Density: 1.30 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004801
CAS Number: 2740-81-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b028d96825ff2c74a638dfa14dd7651f2fcd046&vid=5b028d96825ff2c74a638dfa14dd7651f2fcd046&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR