Isopropyl isothiocyanate ≥98.0%

Supplier: TCI America
Danger

Synonyms: Isopropylisothiocyanate

I0520-5ML I0520-25ML
TCI0520-005MLEA 52 USD
TCI0520-005ML TCI0520-25ML
Isopropyl isothiocyanate ≥98.0%
Isopropyl isothiocyanate

CAS Number: 2253-73-8
MDL Number: MFCD00037830
Molecular Formula: C4H7NS
Molecular Weight: 101.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Flash Point (°C): -15
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Isothiocyanic Acid Isopropyl Ester

Formula: C₄H₇NS
MW: 101.17 g/mol
Boiling Pt: 138 °C
Density: 0.95 (20 °C)
Flash Pt: –15 °C
MDL Number: MFCD00037830
CAS Number: 2253-73-8
EINECS: 218-851-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8619df6e3c5fefeba9775aa58ce330857e4a117f&vid=8619df6e3c5fefeba9775aa58ce330857e4a117f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR