1-Bromo-3-methylbutane ≥96.0%

Supplier: TCI America
Danger

Synonyms: 3-Methylbutylbromide

I0504-500G I0504-25G
TCI0504-500GEA 202.8 USD
TCI0504-500G TCI0504-25G
1-Bromo-3-methylbutane ≥96.0%
1-Bromo-3-methylbutane

CAS Number: 107-82-4
MDL Number: MFCD00000253
Molecular Formula: C5H11Br
Molecular Weight: 151.05
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 21
Specific Gravity (20/20): 1.20


Other Product Names / Synonyms: Isoamyl Bromide

Formula: C₅H₁₁Br
MW: 151.05 g/mol
Boiling Pt: 121 °C
Melting Pt: –112 °C
Density: 1.20 (20 °C)
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00000253
CAS Number: 107-82-4
EINECS: 203-522-9
UN: 2341
ADR: 3,III
Merck Index: 13,05130
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc8f168b3bbc3a68053e11639b9b7fdc9be0d4f5&vid=fd58c834c7fb0e1e545428e130f2ede72df0a978&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR