2-Bromophenyl isocyanate ≥98.0%
Supplier: TCI America
CAS Number: 1592-00-3
MDL Number: MFCD00001995
Molecular Formula: C7H4BrNO
Molecular Weight: 198.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 107
Specific Gravity (20/20): 1.60
Other Product Names / Synonyms: Isocyanic Acid 2-Bromophenyl Ester
Formula:
C₇H₄BrNO Boiling Pt: 62…64 °C (0.5 mmHg) Density: 1.60 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00001995 CAS Number: 1592-00-3 UN: 2206 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...