Dodecyltrimethylammonium chloride 98,0
Supplier: TCI America
|
Warning
|
Synonyms:
(1-Dodecyl)trimethylammonium chloride, N,N,N-Trimethyldodecan-1-aminium chloride, DTAC
I0453-500G
I0453-25G
TCI0453-500GEA
316
USD
TCI0453-500G
TCI0453-025G
Dodecyltrimethylammonium chloride 98,0
Dodecyltrimethylammonium chloride
[Reagent for Ion-Pair Chromatography]
CAS Number: 112-00-5
MDL Number: MFCD00041974
Molecular Formula: C15H34ClN
Molecular Weight: 263.89
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 246
Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Acidic Samples
Other Product Names / Synonyms: IPC-DTMA-Cl, Lauryltrimethylammonium Chloride, Trimethyldodecylammonium Chloride
Formula:
C₁₅H₃₄ClN MW: 263.89 g/mol Melting Pt: 246 °C |
MDL Number:
MFCD00041974 CAS Number: 112-00-5 EINECS: 203-927-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...