4-Isopropoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0432-25G I0432-5G
TCI0432-025GEA 67 USD
TCI0432-025G TCI0432-5G
4-Isopropoxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Isopropoxybenzoic acid

CAS Number: 13205-46-4
MDL Number: MFCD00044318
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 163

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00044318
CAS Number: 13205-46-4
EINECS: 236-169-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bb2fefb374aac5247eac27500d5d9fa75fad273&vid=8bb2fefb374aac5247eac27500d5d9fa75fad273&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR