Ethyl-4-iodobenzoate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
Ethyl 4-iodobenzoate, 4-Iodobenzoic acid ethyl ester
I0369-25G
I0369-5G
TCI0369-025GEA
51
USD
TCI0369-025G
TCI0369-5G
Ethyl-4-iodobenzoate ≥98.0% (by GC)
Ethyl-4-iodobenzoate
CAS Number: 51934-41-9
MDL Number: MFCD00017344
Molecular Formula: C9H9IO2
Molecular Weight: 276.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 283
Flash Point (°C): 110
Specific Gravity (20/20): 1.66
Other Product Names / Synonyms: 4-Iodobenzoic Acid Ethyl Ester
Formula:
C₉H₉IO₂ MW: 276.07 g/mol Boiling Pt: 283 °C Density: 1.66 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00017344 CAS Number: 51934-41-9 EINECS: 429-550-3 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...