(R)-(+)-1-Phenylethyl isocyanate ≥98.0%

Supplier: TCI America

Synonyms: (R)-(+)-α-Phenethyl isocyanate, (R)-(+)-1-Phenethyl isocyanate, (R)-(+)-1-phenylethylisocyanate, (R)-(+)-α-Methylbenzyl isocyanate

I0334-1G I0334-5G
TCI0334-001GEA 43 USD
TCI0334-001G TCI0334-005G TCI0334-25G
(R)-(+)-1-Phenylethyl isocyanate ≥98.0%
(R)-(+)-1-Phenylethyl isocyanate

CAS Number: 33375-06-3
MDL Number: MFCD00063015
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 65
Specific Gravity (20/20): 1.05
Specific rotation [a]20/D: 10 deg (neat)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. Determination (HPLC Labeling Reagents), e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: Isocyanic Acid (R)-(+)-1-Phenylethyl Ester, Isocyanic Acid (R)-(+)-alpha-Methylbenzyl Ester, (R)-(+)-1-Phenylethyl Isocyanate

Formula: C₉H₉NO
Boiling Pt: 74 - 76 °C (7 hPa)
Density: 1.05 (20 °C)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06739387
CAS Number: 33375-06-3
EINECS: 251-485-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e89b893130dab9d5fee1e0204ad7c3bc4d6e881&vid=5e89b893130dab9d5fee1e0204ad7c3bc4d6e881&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR