4-Iodophenol ≥98.0%
Supplier: TCI America
I0311-25G
I0311-10G
TCI0311-025GEA
95
USD
TCI0311-025G
TCI0311-010G
4-Iodophenol ≥98.0%
4-Iodophenol
CAS Number: 540-38-5
MDL Number: MFCD00002327
Molecular Formula: C6H5IO
Molecular Weight: 220.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 91
Formula:
C₆H₅IO MW: 220.01 g/mol Boiling Pt: 287 °C (1013 hPa) Melting Pt: 91 °C Density: 2.13 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002327 CAS Number: 540-38-5 EINECS: 208-745-5 UN: 3261 ADR: 8,III Merck Index: 13,05058 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...