4'-Iodoacetophenone ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-(4-Iodophenyl)ethan-1-one, p-Iodoacetophenone, 1-(4-Iodophenyl)ethanone

I0305-25G I0305-5G
TCI0305-25GEA 71.1 USD
TCI0305-25G TCI0305-005G
4'-Iodoacetophenone ≥98.0%
4'-Iodoacetophenone

CAS Number: 13329-40-3
MDL Number: MFCD00045320
Molecular Formula: C8H7IO
Molecular Weight: 246.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 86

Formula: C₈H₇IO
MW: 246.05 g/mol
Boiling Pt: 145…150 °C (20 mmHg)
Melting Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00045320
CAS Number: 13329-40-3
EINECS: 236-372-8

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR