2-Methoxypropene ≥95.0%
Supplier: TCI America
Synonyms:
2-Methoxy-1-propene, Isopropenyl methyl ether, 2-Methoxyprop-1-ene
I0303-500ML
I0303-25ML
I0303-100ML
TCI0303-500MLEA
822
USD
TCI0303-500ML
TCI0303-25ML
TCI0303-100ML
2-Methoxypropene ≥95.0%
2-Methoxypropene
CAS Number: 116-11-0
MDL Number: MFCD00014929
Molecular Formula: C4H8O
Molecular Weight: 72.11
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 36
Flash Point (°C): -25
Specific Gravity (20/20): 0.77
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Materials Chemistry, Monomers
Other Product Names / Synonyms: Isopropenyl Methyl Ether
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 36 °C Density: 0.77 (20 °C) Flash Pt: –25 °C |
MDL Number:
MFCD00014929 CAS Number: 116-11-0 EINECS: 204-125-3 UN: 1993 ADR: 3,I |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...