Isoamyl alcohol ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol
I0289-500ML
I0289-25ML
TCI0289-500MLEA
49.38
USD
TCI0289-500ML
TCI0289-25ML
Isoamyl alcohol ≥99.0%
Isoamyl alcohol
CAS Number: 123-51-3
MDL Number: MFCD00002934
Molecular Formula: C5H12O
Molecular Weight: 88.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 46
Specific Gravity (20/20): 0.81
Other Product Names / Synonyms: Isoamyl Alcohol, Isobutylcarbinol, Isopentanol, Isopentyl Alcohol
Formula:
(CH₃)₂CHCH₂CH₂OH MW: 88.15 g/mol Boiling Pt: 132 °C Melting Pt: –117 °C Density: 0.81 (20 °C) Flash Pt: 46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002934 CAS Number: 123-51-3 EINECS: 204-633-5 UN: 1105 ADR: 3,III Merck Index: 13,05214 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...