4-Isopropylbenzyl chloride ≥85.0% (contains ca. 10% o-form)
Supplier: TCI America
Synonyms:
1-(Chloromethyl)-4-isopropylbenzene
I0225-500G
I0225-25G
TCI0225-500GEA
585
USD
TCI0225-500G
TCI0225-025G
4-Isopropylbenzyl chloride ≥85.0% (contains ca. 10% o-form)
4-Isopropylbenzyl chloride
CAS Number: 2051-18-5
MDL Number: MFCD00018885
Molecular Formula: C10H13Cl
Molecular Weight: 168.66
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Flash Point (°C): 104
Specific Gravity (20/20): 1.02
Other Product Names / Synonyms: 7-Chloro-p-cymene
Formula:
C₁₀H₁₃Cl MW: 168.67 g/mol Density: 1.02 (20 °C) Flash Pt: 104 °C |
MDL Number:
MFCD00018885 CAS Number: 2051-18-5 EINECS: 218-114-6 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >85.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...