Isovaleric anhydride ≥99.0%

Supplier: TCI America
Danger

Synonyms: Isopentanoic anhydride

I0199-500ML I0199-25ML
TCI0199-500MLEA 413 USD
TCI0199-500ML TCI0199-25ML
Isovaleric anhydride ≥99.0%
Isovaleric anhydride

CAS Number: 1468-39-9
MDL Number: MFCD00015051
Molecular Formula: C10H18O3
Molecular Weight: 186.25
Purity/Analysis Method: >99.0% (T)
Form: Clear Liquid
Boiling point (°C): 215
Flash Point (°C): 91
Specific Gravity (20/20): 0.94

Formula: C₁₀H₁₈O₃
Boiling Pt: 215 °C
Melting Pt: -65 °C
Density: 0.94 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00015051
CAS Number: 1468-39-9
EINECS: 215-994-3
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e17a340d45f79e289dbfd9bdffa425d1dd908e6&vid=8e17a340d45f79e289dbfd9bdffa425d1dd908e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR