Ethyl isovalerate ≥99.0%

Supplier: TCI America

Synonyms: Ethyl isopentanoate, Isovaleric acid ethyl ester, 3-Methylbutyric acid ethyl ester, Isopropyl acetic acid ethyl ester, 3-Methylbutanoic acid ethyl ester

I0194-500ML I0194-25ML
TCI0194-500MLEA 68 USD
TCI0194-500ML TCI0194-025ML
Ethyl isovalerate ≥99.0%
Ethyl isovalerate

CAS Number: 108-64-5
MDL Number: MFCD00009203
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 31
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Isovaleric Acid Ethyl Ester

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 134 °C
Melting Pt: –99 °C
Density: 0.87 (20 °C)
Flash Pt: 31 °C
MDL Number: MFCD00009203
CAS Number: 108-64-5
EINECS: 203-602-3
UN: 3272
ADR: 3,III
Merck Index: 13,03849
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7012c7c8b2680e0b8ab04b838e85ec4915829a01&vid=7012c7c8b2680e0b8ab04b838e85ec4915829a01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR