Phenyl isothiocyanate ≥98.0%

Supplier: TCI America
Danger

I0191-25G I0191-500G I0191-100G
TCI0191-025GEA 25 USD
TCI0191-025G TCI0191-500G TCI0191-100G
Phenyl isothiocyanate ≥98.0%
Phenyl isothiocyanate

CAS Number: 103-72-0
MDL Number: MFCD00004798
Molecular Formula: C7H5NS
Molecular Weight: 135.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 71
Flash Point (°C): 88
Freezing point (°C): -21
Specific Gravity (20/20): 1.14
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Isothiocyanic Acid Phenyl Ester

Formula: C₇H₅NS
MW: 135.19 g/mol
Boiling Pt: 71 °C
Melting Pt: –21 °C
Density: 1.14 (20 °C)
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004798
CAS Number: 103-72-0
EINECS: 203-138-1
UN: 2927
ADR: 6.1 II
Merck Index: 12,07450
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d9dfdf55d3b2bb4203c62229d1fbfebb054a7d0&vid=7d9dfdf55d3b2bb4203c62229d1fbfebb054a7d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR