Isoquinoline ≥95.0%

Supplier: TCI America

I0182-500G I0182-25G
TCI0182-500GEA 147 USD
TCI0182-500G TCI0182-25G
Isoquinoline ≥95.0%
Isoquinoline

CAS Number: 119-65-3
MDL Number: MFCD00006898
Molecular Formula: C9H7N
Molecular Weight: 129.16
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 242
Flash Point (°C): 102
Freezing point (°C): 23

Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 242 °C
Melting Pt: 21 - 26 °C
Density: 1.098 g/cm³ (20 °C)
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006898
CAS Number: 119-65-3
EINECS: 204-341-8
UN: 2811
ADR: 6.1,II
Merck Index: 13,05242
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b410af3ccc5fa5716763526b44a7f5b34554bf16&vid=b410af3ccc5fa5716763526b44a7f5b34554bf16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR