Isopropylamine ≥99.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
2-Aminopropane
I0165-25ML
I0165-500ML
TCI0165-25MLEA
15.67
USD
TCI0165-25ML
TCI0165-500ML
Isopropylamine ≥99.0% (by GC, titration analysis)
Isopropylamine
CAS Number: 75-31-0
MDL Number: MFCD00008082
Molecular Formula: C3H9N
Molecular Weight: 59.11
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 33
Melting point (°C): -95
Flash Point (°C): -20
Specific Gravity (20/20): 0.69
Product Categories: Synthetic Organic Chemistry, Amines (Multi-component Reaction)
Other Product Names / Synonyms: 2-Aminopropane
Formula:
C₃H₉N MW: 59.11 g/mol Boiling Pt: 33 °C Melting Pt: –95 °C Density: 0.69 (20 °C) Flash Pt: –20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008082 CAS Number: 75-31-0 EINECS: 200-860-9 UN: 1221 ADR: 3,I Merck Index: 13,05229 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...