3-Methyl-1-butanethiol ≥94.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-Mercapto-3-methylbutane, 3-Methylbutyl mercaptan, Isopentyl mercaptan

I0149-25ML I0149-250ML
TCI0149-025MLEA 64.11 USD
TCI0149-025ML TCI0149-250ML
3-Methyl-1-butanethiol ≥94.0% (by titrimetric analysis)
3-Methyl-1-butanethiol

CAS Number: 541-31-1
MDL Number: MFCD00004899
Molecular Formula: C5H12S
Molecular Weight: 104.21
Purity/Analysis Method: >94.0% (T)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 19
Specific Gravity (20/20): 0.84


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: Isopentanethiol, 3-Methyl-1-butanethiol

Formula: C₅H₁₂S
MW: 104.22 g/mol
Boiling Pt: 120 °C
Melting Pt: –134 °C
Density: 0.84 (20 °C)
Flash Pt: 19 °C
MDL Number: MFCD00004899
CAS Number: 541-31-1
EINECS: 208-774-3
UN: 1111
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eb1998d4bf10c7235e062f9eabbc83569bdac54&vid=6eb1998d4bf10c7235e062f9eabbc83569bdac54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >94.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR